missing translation for 'onlineSavingsMsg'
Learn More

GL7, PE, clone: GL-7 (GL7), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5902-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15587046

  • £65.34 / 25µg


EU Stock 2
Add to basket

Description

Description

Description: This GL7 monoclonal antibody reacts with a cell-surface protein found on T and B lymphocytes activated in vitro, on bone marrow pre-B-II cells, germinal center B cells, and also human B cell lines Ramos and Daudi. There is strain variability with respect to antigen distribution on thymocytes and Con A-activated spleen cells, with expression in BALB/c greater than that in C57BL/6. GL7 is commonly used as a marker for mouse germinal center B cells. The epitope of GL7 has been identified as a sialic acid glycan moiety called Neu5Ac. This moiety is recognized by CD22.Applications Reported: This GL-7 (GL7) antibody has been reported for use in flow cytometric analysis.Applications Tested: This GL-7 (GL7) antibody has been tested by flow cytometric analyis of stimulated mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

GL7 (T and B Cell Activation Marker)
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
Ly-77, Ly77, B cell activation marker, GL7 antigen
IgM
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Murine
Flow Cytometry
GL-7 (GL7)
PE
Conjugated
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.