missing translation for 'onlineSavingsMsg'
Learn More

GIT 27, Tocris Bioscience™

Immunomodulator; reduces production of pro-inflammatory cytokines

Brand:  Tocris (Bio-Techne) 327010

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13452694

  • £133.77 / 10mg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Orally active immunomodulatory agent that primarily targets macrophages. Inhibits TNF-α secretion via interference of macrophage toll-like receptor (TLR) 4 and TLR 2/6 signaling pathway. Also reduces the secretion of pro-inflammatory cytokines IL1-β, IL-10 and IFN-γ. Antidiabetogenic; prevents IL-β and IFN-γ-induced pancreatic islet cell death in vitro.
Specifications

Chemical Identifiers

6501-72-0
205.213
git 27, git-27, 2-3-phenyl-4,5-dihydro-1,2-oxazol-5-yl acetic acid, 2-3-phenyl-4,5-dihydroisoxazol-5-yl acetic acid, 4,5-dihydro-3-phenyl-5-isoxazoleacetic acid, 3-phenyl-4,5-dihydro-1,2-oxazol-5-yl acetic acid, d03qeb, git27
2-(3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid
C11H11NO3
MUFJHYRCIHHATF-UHFFFAOYSA-N
10798271
C1C(ON=C1C2=CC=CC=C2)CC(=O)O

Specifications

10mg
6501-72-0
git 27, git-27, 2-3-phenyl-4,5-dihydro-1,2-oxazol-5-yl acetic acid, 2-3-phenyl-4,5-dihydroisoxazol-5-yl acetic acid, 4,5-dihydro-3-phenyl-5-isoxazoleacetic acid, 3-phenyl-4,5-dihydro-1,2-oxazol-5-yl acetic acid, d03qeb, git27
C1C(ON=C1C2=CC=CC=C2)CC(=O)O
205.213
205.21
GIT 27
C11H11NO3
MUFJHYRCIHHATF-UHFFFAOYSA-N
2-(3-phenyl-4,5-dihydro-1,2-oxazol-5-yl)acetic acid
10798271
>99%
Videos
Documents