Learn More
GILZ Monoclonal Antibody (CFMKG15), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 14-4033-80
Code : Z2
Additional Details : Weight : 0.01000kg
Description
Description: GILZ is a leucine-zipper-containing protein that is induced by glucocorticoids. It is expressed in mast cells, monocytes, macrophages, dendritic cells, and T cells. GILZ has been shown to inhibit production of IL-2 as well as TCR-driven upregulation of FasL. GILZ mediates these changes in transcription, at least in part, by inhibiting DNA-binding of the transcription factors AP-1 and NF-kappaB. GILZ has also been shown to mediate anti-inflammatory signals in macrophages and dendritic cells by interfering with NF-kappaB-mediated signaling. Although GILZ has been reported to localize to the nucleus, more recent reports as well as testing at eBioscience have observed predomitly cytoplasmic staining. Applications Reported: This CFMKG15 antibody has been reported for use in intracellular staining followed by flow cytometric analysis, immunoblotting (WB), and immunocytochemistry. (Fluorochrome conjugated CFMKG15 is recommended for use in intracellular flow cytometry.). Applications Tested: This CFMKG15 antibody has been tested by western blot analysis of dexamethasone-treated mouse thymocytes. CFMKG15 recognizes endogenous mouse GILZ with an approximate molecular weight of 14 kDa; a non-specific band of approximately 95 kDa is also observed. CFMKG15 also recognizes endogenous human GILZ in dexamethasone-treated peripheral blood mononuclear cells, with an approximate molecular weight of 14 kDa; non-specific bands may also appear at approximately 33 kDa and 95 kDa.
This can be used at less than or equal to 2 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered. The TSC22 domain family member 3 protein (TSC22D3) is a leucine zipper protein that functions as a transcriptional regulator. The expression of TSC22D3 is stimulated by glucocorticoids and IL-10 and is thought to play a key role in the anti-inflammatory and immunosuppressive effects of these molecules. TSC22D3 can physically interact with and inhibit the activities of key inflammatory signaling mediators such NF-Kappa-B and AP-1. TSC22D3 functions as a transcriptional co-activator for various nuclear receptors and NF-Kappa-B. It has also been shown to be involved in the differentiation of mesenchymal stem cells towards osteoblasts and bone formation.Specifications
GILZ | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q99576, Q9Z2S7 | |
TSC22D3 | |
Affinity chromatography | |
RUO | |
14605, 1831 | |
4° C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
CFMKG15 | |
Unconjugated | |
TSC22D3 | |
TSC22D3, TSC22 domain family member 3, TSC-22 related protein, delta sleep inducing peptide, DIP, DSIPI, GILZ, TSC-22R, DSIP-immunoreactive leucine zipper protein | |
Rat | |
25 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.