Learn More
Gilson™ Safe Aspiration Station
Vacuum-Controlled Aspiration System with small footprint
Brand:  Gilson™ F110742
Code : 50
Additional Details : Weight : 1.10000kg
 View more versions of this product
Product Code. 12376162
  • £302.00 / Each
Call for latest availability
+44 (0)1509 555 500
Add to basket
Description
Description
Closed, stand-alone unit designed to prevent contamination hazards.
  • Compact design with adjustable speed allowing quick aspiration
  • Always ready to use by simply pressing the handle button
  • Handle accommodates different types of adaptors to aspirate from a variety of vessels
  • Convenient coupling for quick bottle emptying
  • In-line hydrophobic filter traps aerosols
  • Connect two handles to the station with the Y-shape connector
  • Motor starts only when the vacuum pressure drops
  • Level detection prevents overflow
Specifications
Specifications
Safe Aspiration Kit
1
Videos