missing translation for 'onlineSavingsMsg'
Learn More

Gilson™ PIPETMAN™ Starter Kit

Contains boxes of certified-quality PIPETMAN™ Tips and useful accessories. Gilson™ PIPETMAN™ Starter Kit includes single pipet holders, COLORIS™ Identification Clips (mixed colors), and a Gilson Guide to Pipetting (except Large-volume Kit).

Brand:  Gilson™ F167400

Code : 50

Additional Details : Weight : 0.60000kg

 View more versions of this product

Product Code. 10797404

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Contains boxes of certified-quality PIPETMAN Tips and useful accessories.
  • Single pipet holders
  • COLORIS™ Identification Clips (mixed colors)
  • Gilson Guide to Pipetting (except Large-volume Kit)
Specifications

Specifications

Manual Air Displacement
Reusable
Ergonomic
1
Non-sterile
500 μL to 10 mL
P5000, P10mL pipet, one package of filters and tip for each pipet (P5000: 10 filters and 5 tips, P10mL: 10 filters and 2 tips)
14.37 in.
12.01 in.
4.72 in.
Certified Quality
Blue/White
Manual Air Displacement Pipette
Polypropylene
Pipetman™
Pipette Starter Kit
Non-Filtered
3 Years
36.5 cm
30.5 cm
12 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers