Learn More
Gilson™ PIPETMAN™ Diamond Blister Pack Refill Pipette Tips
Autoclavable or gamma-ray sterlized
£70.30 - £198.00
Product Code | Brand | Volume (Metric) | Filtered | Sterility | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Filtered | Sterility | Price | Quantity & Availability | |||
15247783
|
Gilson™
F172511 |
100 to 1000 μL | Non-Filtered | Sterile |
£79.05
Pack of 960 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
15237783
|
Gilson™
F172510 |
100 to 1000 μL | Non-Filtered | Non-sterile |
£70.30
Pack of 960 |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
15707627
|
Gilson™
F172313 |
2 to 30 μL | Filtered | Sterile |
£167.00
Pack of 960 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
15227783
|
Gilson™
F172311 |
2 to 200 μL | Non-Filtered | Sterile |
£80.80
Pack of 960 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||
15267783
|
Gilson™
F172613 |
100 to 1000 μL | Filtered | Sterile |
£198.00
Pack of 960 |
Estimated Shipment: 20-06-2024
Log in to see stock. |
|||
15257783
|
Gilson™
F172513 |
2 to 200 μL | Filtered | Sterile |
£167.00
Pack of 960 |
Estimated Shipment: 10-07-2024
Log in to see stock. |
|||
15297773
|
Gilson™
F172211 |
0.1 to 20 μL | Non-Filtered | Sterile |
£88.85
Pack of 960 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||
15217783
|
Gilson™
F172310 |
2 to 200 μL | Non-Filtered | Non-sterile |
£73.20
Pack of 960 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
15207783
|
Gilson™
F172213 |
0.1 to 10 μL | Filtered | Sterile |
£185.00
Pack of 960 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
|||
15287773
|
Gilson™
F172210 |
0.1 to 20 μL | Non-Filtered | Non-sterile |
£79.05
Pack of 960 |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||
Description
- Color-coded tip packaging corresponds to industry standards for volume color matching to pipettes
- Features graduations at varying volumes allowing for quick-checks when aspirating samples
- Standard or filtered tips
- Validated for use with 384-well tips: F172211G, F172311G and F17511G
Specifications
Clear | |
Pipetman Diamond | |
Polypropylene |