missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ PIPETMAN™ Diamond™ Tips Blister Refill
Individually wrapped racks of tips that are convenient, clean and eco-friendly
£43.05 - £49.95
Specifications
Volume (Metric) | 20 to 300 μL |
---|---|
Color | Clear |
Model | D300; Standard |
Length (Metric) | 5.1cm |
For Use With (Equipment) | Pipetmax |
Product Code | Brand | Autoclavable | Sterility | Model | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Autoclavable | Sterility | Model | Price | Quantity & Availability | |||
15374775
|
Gilson™
F172400 |
Autoclavable | Non-sterile | D300; Standard |
£43.05
Pack of 960 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
15384775
|
Gilson™
F172401 |
Not Autoclavable | Sterile | D300; Standard |
£49.95
Pack of 960 |
Estimated Shipment: 03-09-2024
Log in to see stock. |
|||
Description
- Color-coded racks to indicate tip models and PIPETMAN model compatibility
- Available as autoclavable or gamma-ray sterilized
- Fit with PIPETMAX (special tips validated for use with 384 well plates)
- 10 individually wrapped racks of 96 tips
- Ready to use
Specifications
20 to 300 μL | |
D300; Standard | |
Pipetmax | |
Polypropylene |
Clear | |
5.1cm | |
Pipetman Diamond |