missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gilson™ MACROMAN™ Motorized Pipette Controller Accessory, Filter
For use with MACROMAN Pipette Controller
£15.65 - £28.20
Specifications
For Use With (Equipment) | Macroman pipette controller |
---|---|
Quantity | 1/Cs. |
Warranty | 1 Year |
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
12326162
|
Gilson™
F1077503 |
Standard Filter, 0.20 μm |
£15.65
Each |
Estimated Shipment: 07-08-2024
Log in to see stock. |
|||||
12346162
|
Gilson™
F1077525 |
Sterile Filter, 0.45 μm |
£28.20
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
10231220
|
Gilson™
F1077526 |
Sterile Filter, 0.20 μm |
£28.20
Each |
Estimated Shipment: 28-06-2024
Log in to see stock. |
|||||
Description
- Robust and reliable pipette controller system
- For use with glass and plastic pipettes
Specifications
Macroman pipette controller | |
1/Cs. | |
1 Year |