Learn More
Gilson™ Accessory for PIPETMAN™ Complete Pipetting System - Reagent Reservoir
Ideal for dispensing reagents with multichannel pipettes. Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Reagent Reservoirs' unique shape allows for efficient and nearly complete sample withdrawal.
Brand: Gilson™ F267670
Code : 50
Additional Details : Weight : 0.92700kg
Description
- Angled sides and V-shaped bottoms
- Pour-off spouts on all corners
- Graduations on inside walls at middle volume
Specifications
White | |
Polystyrene | |
PIPETMAN™ Complete Pipetting System |
All Pipettes with Tips Mounted | |
50 mL | |
100 Pk. |