Learn More
Gilson™ Accessory for PIPETMAN™ Complete Pipetting System- Coloris™ Colored Identifi cation Clips
GLP-friendly, brightly colored clips made of polyacetal are used for identifying Pipetteman™ and Neo™ pipets
Brand:  Gilson™ F161306
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10337662
  • £16.80 / Pack of 10
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
Description
Description
  • Brightly colored clips
  • Made of polyacetal
  • Used for identifying Pipetteman and Neo pipettes
Specifications
Specifications
White
Polyacetal
Pipetteman™ and Neo™ pipettes