missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ VEGF Recombinant Rat Protein
Recombinant VEGF is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Exchange Chromatography |
---|---|
Gene Alias | VEGF |
Gene ID (Entrez) | 83785 |
Description
VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.Approved for shipment on Wet or Dry Ice.
Specifications
Exchange Chromatography | |
VEGF | |
83785 |
For Research Use Only. Not for use in diagnostic procedures.