missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ VEGF Recombinant Rat Protein

Recombinant VEGF is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PRG0115

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10672274

  • £279.46 / Each


Estimated Shipment
09-07-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis
83785
Exchange Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
38.4kDa
95%
Rat
Carrier Protein
VEGF (Vascular Endothelial Growth Factor)
VEGF
Growth Factors & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.