missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ VEGF Recombinant Mouse Protein

Recombinant Mouse Vascular Endothelial Cell Growth Factor (VEGF) is a bioactive protein intended for use in cell culture applications

£238.00 - £3660.00

Specifications

Purity 95%
Purification Method Exchange Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias VEGF
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10232473 View Documents Gibco™
PMG0114
10μg £238.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Please to purchase this item. Need a web account? Register with us today!
10561645 View Documents Gibco™
PMG0111
100μg £1018.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Please to purchase this item. Need a web account? Register with us today!
10028912 View Documents Gibco™
PMG0113
1mg £3660.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 13-03-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human umbilical vein cells (HUVEC)

Approved for shipment on Wet or Dry Ice.

Specifications

Specifications

95%
<0.1ng/μg
VEGF
Exchange Chromatography
Lyophilized
22339

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.