missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ VEGF Recombinant Mouse Protein

Recombinant Mouse Vascular Endothelial Cell Growth Factor (VEGF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMG0113

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10028912

  • £3660.00 / Each

Estimated Shipment date
from Supplier 06-03-2020

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

VEGF exerts angiogenic, mitogenic and vascular permeability-enhancing activities specific for endothelial cells. VEGF has also been shown to be chemotactic for monocytes and osteoblasts.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human umbilical vein cells (HUVEC)

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis
22339
Exchange Chromatography
1mg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
38.8kDa
95%
Mouse
Carrier Protein
VEGF (Vascular Endothelial Growth Factor)
VEGF
Growth Factors & Receptors
Videos
Documents

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.