Learn More
Gibco™ Human TRAIL (TNFSF10) Recombinant Protein
Human TRAIL (TNFSF10) Recombinant Protein
Brand: Gibco™ PHC1634
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human TRAIL (TNFSF10) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
TRAIL preferentially induces apoptosis in transformed and tumor cells, but does not appear to kill normal cells although it is expressed at a significant level in most normal tissues. The binding of this protein to its receptors has been shown to trigger the activation of MAPK8/JNK, caspase 8, and caspase 3.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
19.6 kDa | |
50 ug | |
168 aa | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Apoptosis, Oncology, Signal Transduction | |
98 % |
8743 | |
Sequential Chromatography | |
TRAIL | |
Human | |
Carrier-Free | |
50 μg | |
E. coli | |
Recombinant, Ligand | |
Other Proteins | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.