missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ TPO Recombinant Mouse Protein
Recombinant TPO is a bioactive protein intended for use in cell culture applications
£216.00 - £986.00
Specifications
Purity | 95 % |
---|---|
Purification Method | Sequential Chromatography |
Gene Alias | TPO |
Gene ID (Entrez) | 22018 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10334123
|
Gibco™
PMC1144 |
10μg |
£216.00
Each |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10009422
|
Gibco™
PMC1145 |
25μg |
£287.00
Each |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10161203
|
Gibco™
PMC1141 |
100μg |
£986.00
Each |
Estimated Shipment 22-03-2021 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.Approved for shipment on Wet or Dry Ice.
Specifications
95 % | |
Sequential Chromatography | |
TPO | |
22018 |
For Research Use Only. Not for use in diagnostic procedures.