Learn More
Gibco™ TPO Recombinant Mouse Protein
Recombinant TPO is a bioactive protein intended for use in cell culture applications
Brand:  Gibco™ PMC1145
Code : 50
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10009422
  • £287.00 / Each
Estimated Shipment: 30-09-2021
For Research Use Only. All usage must comply with product instructions.
Description
Description
TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Specifications
< 0.1 ng/µg
Approved for shipment on Wet or Dry Ice
Lyophilized
Growth Factors & Receptors
TPO
Endocrinology, Stem Cell Research, Angiogenesis
Cell Culture
17 kDa
95 %
TPO
25 µg
E. coli
Gibco™
Recombinant, Ligand
Carrier-Free
Mouse
22018
Sequential Chromatography
25μg
Yes