missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TPO Recombinant Mouse Protein

Recombinant TPO is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC1141

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10161203

  • £921.72 / Each


Estimated Shipment
14-05-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

TPO is a humoral growth factor that is necessary for megakaryocyte proliferation and maturation, as well as for thrombopoiesis. This protein is the ligand for MLP/C_MPL, the product of myeloproliferative leukemia virus oncogene.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis; Cell Proliferation; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
17kDa
95%
Mouse
Carrier Protein
TPO
TPO
Growth Factors & Receptors
Lyophilized
Sequential Chromatography
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.