missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TNF Recombinant Rat Protein

Recombinant TNF is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PRC3013

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10282973

  • £1952.88 / Each


Estimated Shipment
11-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Signaling
24835
Sequential Chromatography
1mg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
17kDa
95%
Rat
Carrier Protein
TNF (Tumor Necrosis Factor)
TNF-α
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.