missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TNF Recombinant Mouse Protein

Recombinant Mouse Tumor Necrosis Factor-α (TNF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC3014

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10620924

  • £78.12 / Each


EU Stock 3
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent cytotoxicity of L929 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Cell Signaling
21926
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
17.5kDa
95%
Mouse
Carrier Protein
TNF (Tumor Necrosis Factor)
TNF-α
Cytokines & Receptors
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.