Learn More
Gibco™ TNF Recombinant Mouse Protein
Recombinant Mouse Tumor Necrosis Factor-α (TNF) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PMC3015
Code : 50
Additional Details : Weight : 0.02100kg
Description
TNF causes cytolysis and cytostasis of many tumor cell lines. TNF has a wide spectrum of activities, including chemotaxis of neutrophils, alteration of the endothelium, inhibition of anticoagulatory mechanisms, and promotion of angiogenesis.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent cytotoxicity of L929 cells
Shipping Condition: Wet Ice.
Specifications
Store in refrigerator (2–8°C). | |
100 µg | |
Carrier-Free | |
E. coli | |
Cytokines & Receptors | |
TNF (Tumor Necrosis Factor) | |
Mouse | |
21926 | |
Sequential Chromatography | |
100μg |
< 0.1 ng/µg | |
Gibco™ | |
Wet Ice | |
Lyophilized | |
Recombinant, Ligand | |
Immunology, Signal Transduction, Stem Cell Research, Inflammation, Oncology | |
Cell Culture | |
17.5 kDa | |
95 % | |
TNF-α |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.