missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TIMP1 Recombinant Human Protein

Recombinant TIMP1 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC8021

UNSPSC : 12352202

Code : 50

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10135794

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

TIMP1 is an inhibitor of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix. In addition to its inhibitory role against most of the knownmMPs, the encoded protein is able to promote cell proliferation in a wide range of cell types, and may also have an anti-apoptotic function.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
100μg
Store at 2° to 8°C.
TIMP
Proteases, Peptidases & Inhibitors
443331, 73858576
Human
Recombinant
Cell Culture