missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ TGFB2 Recombinant Human Protein

Recombinant Human Transforming Growth Factor β 2 (TGFB2) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHG9124

Code : 50

Additional Details : Weight : 0.00001kg

 View more versions of this product

Product Code. 11340862

  • £417.15 / Each


Estimated Shipment
12-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

TGFB2 binds the transforming growth factor-β receptor and plays a role in regulation of cell growth and proliferation. It may be involved in mesenchymal-epithelial cell interactions during development.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High purity recombinant bioactive protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High biological activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by its ability to inhibit the mouse IL-4 dependent proliferation of mouse HT-2 cells

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Stem Cell Research; Stem Cell Culture; Stem Cell Differentiation; Embryonic Stem Cell Culture; Hematopoietic Stem Cell Culture; Mesenchymal Stem Cell Culture; Differentiation & Development
Sequential Chromatography
10μg
Store at -5° to -30°C.
Recombinant/Ligand
<1 EU/mL
Inhibition of proliferation of HT2 cells
Growth Factors & Receptors
112 aa
25kDa
95%
Human
Carrier Protein
TGFs (Transforming Growth Factors)
TGF-β2
Cell Culture
Mammalian
1ng/mL
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.