Learn More
Gibco™ SDF1B Recombinant Human Protein
Recombinant SDF1B is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC1364
Code : 50
Additional Details : Weight : 0.50000kg
Description
SDF1B is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture; Mammalian Cell Culture | |
6387 | |
Sequential Chromatography | |
10μg | |
Store at 2° to 8°C. | |
Recombinant/Ligand | |
<0.1ng/μg | |
Cell Culture | |
E. coli |
Lyophilized | |
8.7kDa | |
95% | |
Human | |
Carrier Protein | |
SDFs (Stromal Cell-Derived Factors) | |
SDF-1β | |
Chemokines & Receptors |
For Research Use Only. Not for use in diagnostic procedures.