missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ SDF1A Recombinant Human Protein
Recombinant SDF1A is a bioactive protein intended for use in cell culture applications
£522.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 6387 |
Molecular Weight (g/mol) | 8 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | SDF-1α |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10120173
|
Gibco™
PHC1346 |
25μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
SDF1A is a small cytokine that belongs to the intercrine family, members of which activate leukocytes and are often induced by proinflammatory stimuli such as lipopolysaccharide, TNF, or IL-1.Approved for shipment on Wet or Dry Ice.
Specifications
Cell Culture | |
8 kDa | |
SDF-1α | |
Recombinant | |
Carrier-Free | |
Approved for shipment on Wet or Dry Ice | |
Chemokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Cardiovascular Research, Neurobiology | |
95 % |
6387 | |
Sequential Chromatography | |
Human | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
SDFs (Stromal Cell-Derived Factors) | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.