missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ SCF (C-Kit Ligand) Recombinant Mouse Protein

Recombinant Mouse Stem Cell Factor (SCF, C-Kit Ligand) is a bioactive protein intended for use in cell culture applications

£153.00 - £2230.00

Specifications

Purity 95%
Purification Method Sequential Chromatography
Endotoxin Concentration <0.1ng/μg
Format Lyophilized
Gene Alias SCF, c-kit Ligand
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10503205 View Documents Gibco™
PMC2114
10μg £153.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 04-03-2020
Please to purchase this item. Need a web account? Register with us today!
10009462 View Documents Gibco™
PMC2115
25μg £292.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 03-03-2020
Please to purchase this item. Need a web account? Register with us today!
10160813 View Documents Gibco™
PMC2111
100μg £1012.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 10-03-2020
Please to purchase this item. Need a web account? Register with us today!
10622264 View Documents Gibco™
PMC2113L
1mg £2230.00
Each View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 10-03-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SCF is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of human MO-7e cells

Shipping Condition: Wet Ice.

Specifications

Specifications

95%
<0.1ng/μg
SCF, c-kit Ligand
Sequential Chromatography
Lyophilized
17311

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.