missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ RANTES Recombinant Human Protein
Recombinant RANTES is a bioactive protein intended for use in cell culture applications
£76.50
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 6352 |
Molecular Weight (g/mol) | 7.8 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | RANTES |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10079352
|
Gibco™
PHC1054 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.Shipping Condition: Dry Ice.
Specifications
Cell Culture | |
7.8 kDa | |
RANTES | |
Recombinant | |
Carrier-Free | |
Wet Ice | |
Chemokines & Receptors | |
Lyophilized | |
Immunology, Stem Cell Research, Oncology | |
95 % |
6352 | |
Sequential Chromatography | |
Human | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
Rantes | |
Gibco™ |
For Research Use Only. Not for use in diagnostic procedures.