missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ RANTES Recombinant Human Protein

Recombinant RANTES is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PHC1051

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10583785

  • £470.71 / Each


Estimated Shipment
15-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

RANTES functions as a chemoattractant for blood monocytes, memory T helper cells and eosinophils. It causes the release of histamine from basophils and activates eosinophils. RANTES is one of the major HIV-suppressive factors produced by CD8+ cells. It functions as one of the natural ligands for the chemokine receptor CCR5 and it suppresses in vitro replication of the R5 strains of HIV-1, which use CCR5 as a coreceptor.

Shipping Condition: Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
6352
Sequential Chromatography
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
7.8kDa
95%
Human
Carrier Protein
Rantes
RANTES
Chemokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.