missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ PDGF-BB Recombinant Mouse Protein

Recombinant Mouse Platelet Derived Growth Factor-BB (PDGF-BB) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMG0045

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10502635

  • £285.65 / Each


Estimated Shipment
09-07-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose-dependent proliferation of BALB/c 3T3 cells

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
18591
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
25kDa
95%
Mouse
Carrier Protein
PDGFs (Platelet-Derived Growth Factors)
PDGF-BB
Growth Factors & Receptors
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.