Learn More
Gibco™ Human PDGF-BB Recombinant Protein
Recombinant Human Platelet Derived Growth Factor-BB (PDGF-BB) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHG0045
Code : 50
Additional Details : Weight : 0.02100kg
Description
Human PDGF-BB Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
Members of the PDGF family are mitogenic factors for cells of mesenchymal origin.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of BALB/c 3T3 cells
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
25.4 kDa | |
10μg | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Full Length, Recombinant, Ligand | |
PDGFs (Platelet-Derived Growth Factors) | |
Gibco™ |
5155 | |
Sequential Chromatography | |
PDGF-BB | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Growth Factors & Receptors | |
Lyophilized | |
Immunology, Neurobiology, Stem Cell Research, Angiogenesis, Oncology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.