missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Gentamicin (50 mg/mL)

Water-soluble antibiotic drug originally purified from the fungus Micromonospora purpurea

Brand:  Gibco™ 15750078

Code : NEW

Additional Details : Weight : 1.00000kg

 View more versions of this product

Product Code. 15400604

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

  • Gentamicin sulfate is a water-soluble antibiotic originally purified from the fungus Micromonospora purpurea
  • It acts by binding to the 30S subunit of the bacterial ribosome leading to inhibition of protein synthesis and death in susceptible bacteria
  • Gibco™ Gentamicin is effective against a wide variety of gram-positive and gram-negative bacteria, and is used to prevent the contamination of cell cultures by bacteria
  • The recommended working concentration ranges from 0.5 to 50 μg /mL
  • Store at 15°C to 30°C
Specifications

Specifications

Gentamicin
Yes
10 x 10 mL
Room Temperature
Liquid
50 mg/mL
Prevention of Cell Culture Contamination
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.