missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ MCP-1 (MCAF) Recombinant Rat Protein
Recombinant MCP-1 (MCAF) is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Sequential Chromatography |
---|---|
Gene Alias | MCP-1 (MCAF) |
Gene ID (Entrez) | 24770 |
Description
MCP-1 is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils.Shipping Condition: Wet Ice.
Specifications
Sequential Chromatography | |
MCP-1 (MCAF) | |
24770 |
For Research Use Only. Not for use in diagnostic procedures.