missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MCP-1 (MCAF) Recombinant Rat Protein

Recombinant MCP-1 (MCAF) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PRC1011

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10485903

  • £913.47 / Each


Estimated Shipment
15-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

MCP-1 is structurally related to the CXC subfamily of cytokines. Members of this subfamily are characterized by two cysteines separated by a single amino acid. This cytokine displays chemotactic activity for monocytes and basophils but not for neutrophils or eosinophils.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
24770
Sequential Chromatography
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
14kDa
95%
Rat
Carrier Protein
MCP/MCAF Chemokines
MCP-1, MCAF
Chemokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.