missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IP10 Recombinant Mouse Protein

Recombinant IP10 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC1083

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10447253

  • £1888.80 / Each


Estimated Shipment
13-03-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
8.7kDa
95%
Mouse
Carrier Protein
IP-10
IP-10
Chemokines & Receptors
Lyophilized
Sequential Chromatography
1mg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Videos