missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ IP10 Recombinant Human Protein
Recombinant IP10 is a bioactive protein intended for use in cell culture applications
POA
Specifications
Purification Method | Sequential Chromatography |
---|---|
Gene Alias | IP-10 |
Gene ID (Entrez) | 3627 |
Description
IP10 is a chemokine of the CXC subfamily and ligand for the receptor CXCR3. Binding of this protein to CXCR3 results in pleiotropic effects, including stimulation of monocytes, natural killer and T-cell migration, and modulation of adhesion molecule expression.Approved for shipment on Wet or Dry Ice.
Specifications
Sequential Chromatography | |
IP-10 | |
3627 |
For Research Use Only. Not for use in diagnostic procedures.