missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein
Recombinant IL8 Monocyte is a bioactive protein intended for use in cell culture applications
£192.00 - £2610.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 3576 |
Gene Alias | IL-8 Monocyte |
Species | Human |
Content And Storage | Store in refrigerator (2–8°C). |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10272833
|
Gibco™
PHC0884 |
25μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10282833
|
Gibco™
PHC0885 |
50μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10477063
|
Gibco™
PHC0881 |
100μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10202553
|
Gibco™
PHC0883 |
1mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Human IL-8 (CXCL8) (Monocyte-derived) Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL8 is chemotactic for all known types of migratory immune cells. IL8 also inhibits the adhesion of leukocytes to activated endothelial cells.Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
IL-8 Monocyte | |
Store in refrigerator (2–8°C). | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
3576 | |
Human | |
Wet Ice | |
Chemokines & Receptors | |
Lyophilized | |
Immunology, Angiogenesis, Respiratory Research, Virology, Stem Cell Research, Inflammation |
For Research Use Only. Not for use in diagnostic procedures.