missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL7 Recombinant Mouse Protein

Recombinant IL7 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0075

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10120873

  • £88.95 / Each


Estimated Shipment
05-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Cell Proliferation; Stem Cell Research; Stem Cell Culture; Hematopoietic Stem Cell Culture
15kDa
95%
Mouse
Carrier Protein
Interleukins
IL-7
Cytokines & Receptors
Lyophilized
Sequential Chromatography
5μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Videos

For Research Use Only. Not for use in diagnostic procedures.