Learn More
Gibco™ IL7 Recombinant Mouse Protein
Recombinant IL7 is a bioactive protein intended for use in cell culture applications
Brand:  Gibco™ PMC0071
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10079172
  • £1460.00 / Each
Estimated Shipment: 07-02-2022
For Research Use Only. All usage must comply with product instructions.
Description
Description
IL7 stimulates the proliferation of pre-B and pro-B cells. IL7 also supports the maturation of megakaryocytes.
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Specifications
< 0.1 ng/µg
Approved for shipment on Wet or Dry Ice
Lyophilized
Cytokines & Receptors
Interleukins
Immunology, Signal Transduction, Stem Cell Research, Drug Development, Oncology
Cell Culture
Sequential Chromatography
100μg
100 µg
E. coli
Gibco™
Recombinant, Ligand
Carrier-Free
Mouse
15 kDa
95 %
IL-7