Learn More
Gibco™ Human IL-5 Recombinant Protein
Human IL-5 Recombinant Protein
Brand: Gibco™ PHC0055
Code : 50
Additional Details : Weight : 0.05250kg
Description
Human IL-5 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL5 promotes the growth and differentiation of eosinophils.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the IL5 dose dependent proliferation of TF-1 cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Approved for shipment at Room Temperature or on Wet or Dry Ice.
Specifications
Cell Culture | |
26 kDa | |
10 ug | |
232 aa each (homodimer) | |
Store in freezer (-5 to -30°C). | |
< 0.1 ng/μg | |
Approved for shipment on Wet or Dry Ice | |
Cytokines & Receptors | |
Lyophilized | |
Respiratory Research, Immunology | |
98 % |
3567 | |
Sequential Chromatography | |
IL-5 | |
Human | |
Carrier-Free | |
10 μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.