missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL4 Recombinant Swine Protein

Recombinant IL4 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PSC0044

Code : 50

Additional Details : Weight : 0.05250kg

 View more versions of this product

Product Code. 10436093

  • £213.21 / Each


Estimated Shipment
15-06-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
397225
Sequential Chromatography
10μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
12.5kDa
95%
Swine/Porcine
Carrier Protein
Interleukins
IL-4
Cytokines & Receptors
Videos

For Research Use Only. Not for use in diagnostic procedures.