missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL4 Recombinant Swine Protein

Recombinant IL4 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PSC0041

UNSPSC : 12352202

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10435903

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.

Approved for shipment on Wet or Dry Ice.

Description & Specifications

Specifications

Cell Culture; Mammalian Cell Culture; Cell Analysis; Cell Viability, Proliferation & Function; Angiogenesis; Cell Proliferation
12.5kDa
95%
Swine/Porcine
Carrier Protein
Interleukins
IL-4
Cytokines & Receptors
Lyophilized
Sequential Chromatography
100μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli