missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL4 Recombinant Mouse Protein

Recombinant Mouse Interleukin 4 (IL4) is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC0043

Code : 50

Additional Details : Weight : 0.99000kg

 View more versions of this product

Product Code. 10358369

  • £4604.10 / Each


Estimated Shipment
05-07-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL4 promotes the proliferation and differentiation of activated B cells. In addition, IL4 up-regulates class II MHC antigen expression and IgE receptors.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by measuring the dose-dependent proliferation of murine HT-2 cells

Approved for shipment at Room Temperature or on Wet or Dry Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
10946583, 3565
Sequential Chromatography
1mg
Store at -5° to -30°C.
Interleukins
Cell Culture
E. coli
Lyophilized
14kDa
95%
Mouse
Recombinant/Ligand
IL-4
Cytokines & Receptors
Videos

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.