missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL33 Recombinant Mouse Protein

Recombinant IL33 is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PMC4044

Code : 50

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 10627615

  • £215.53 / Each


Estimated Shipment
05-07-2018
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL33 is a proinflammatory cytokine with structural homology similar with IL-1 family cytokines. IL33 exerts its biological effects by binding to the receptor ST2 and forming a complex with IL-1 Receptor Accessory Protein. Intracellular proteins within in the NFκB and MAP kinase signaling pathways are activated leading to the production of the type 2 cytokines IL-5 from polarized TH2 cells.

Shipping Condition: Wet Ice.

Specifications

Specifications

NM_006853
Lyophilized
18kDa
95%
Mouse
Carrier Protein
Interleukins
IL-33
Cell Culture
E. coli
1.0 x 108 - 1.0 x 107 units/mg
Cell Culture; Mammalian Cell Culture
IL33
PAGE
10μg
Lyophilized Mouse IL-33 should be stored at 2° to 8°C, preferably desiccated. Store reconstituted Mouse IL-33 at ≤ –20°C. Avoid repeated freeze/thaw cycles.
Recombinant/Ligand
<0.1ng/μg
T Cells
Cytokines & Receptors
267 aa
Mouse
Videos

For Research Use Only. Not for use in diagnostic procedures.