missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human IL-23 Recombinant Protein
Recombinant Human IL23 is a bioactive protein intended for use in cell culture applications
£354.00 - £2065.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Molecular Weight (g/mol) | 55 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-23 |
Protein Length | 170 aa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11390822
|
Gibco™
PHC9324 |
10μg |
£354.00
Each |
Estimated Shipment: 11-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11300832
|
Gibco™
PHC9321 |
100μg |
£2065.00
Each |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Human IL-23 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL23 is important in the inflammatory response. IL23 induces the secretion of IFN-γ, and stimulates the proliferation of Human T cells. Recombinant Human IL23 is one of the many Gibco recombinant bioactive proteins available from Life Technologies.- High purity: minimal interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, and protease assays; specific activity is determined by measuring the IL23 dose dependent secretion of IL-17 from mouse splenocytes
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
Sequential Chromatography | |
170 aa | |
2° to 8° C | |
0.3 ng/ml | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Inflammation, Signal Transduction | |
95 % |
55 kDa | |
IL-23 | |
Human | |
Carrier-Free | |
< 1 EU/ml | |
Mammalian | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.