missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse IL-2 Recombinant Protein
Recombinant IL-2 is a bioactive protein intended for use in cell culture applications
£71.75 - £2640.00
Specifications
For Use With (Application) | Cell Culture |
---|---|
Gene ID (Entrez) | 16183 |
Molecular Weight (g/mol) | ∼17 kDa |
Purification Method | Sequential Chromatography |
Gene Alias | IL-2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10374733
|
Gibco™
PMC0024 |
10μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10662264
|
Gibco™
PMC0025 |
25μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10202543
|
Gibco™
PMC0021 |
100μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10610734
|
Gibco™
PMC0023 |
1mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Mouse IL-2 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL-2 causes proliferation of T cells and is a central regulator of immune responses.Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
∼17 kDa | |
IL-2 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
16183 | |
Sequential Chromatography | |
Mouse | |
Carrier-Free | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Angiogenesis, Oncology | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.