Learn More
Gibco™ Mouse IL-2 Recombinant Protein
Recombinant IL-2 is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PMC0023
Code : 50
Additional Details : Weight : 0.50000kg
Description
Mouse IL-2 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL-2 causes proliferation of T cells and is a central regulator of immune responses.Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
∼17 kDa | |
1mg | |
Mouse | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand | |
Interleukins | |
Gibco™ |
16183 | |
Sequential Chromatography | |
IL-2 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Angiogenesis, Oncology | |
95 % |
For Research Use Only. Not for use in diagnostic procedures.