Learn More
Gibco™ Human IL-2 Recombinant Protein
Recombinant Human Interleukin 2 (IL2) is a bioactive protein intended for use in cell culture applications
Brand: Gibco™ PHC0023
Code : 50
Additional Details : Weight : 0.50000kg
Description
Human IL-2 Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN
IL2 causes proliferation of T-cells and is a central regulator of immune responses.- High purity: no interference from other proteins or contaminants
- High biological activity: more results with less protein
- Proven compatibility: Gibco proteins bioassayed with Gibco media
- High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
- High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of mouse CTLL-2 cells
- Proven Compatibility: Gibco proteins are bioassayed with Gibco media
Shipping Condition: Wet Ice.
Specifications
Cell Culture | |
15.5 kDa | |
1mg | |
Human | |
Carrier-Free | |
1 mg | |
E. coli | |
Recombinant, Ligand, Mutant | |
Interleukins | |
Gibco™ |
3558 | |
Membrane-Filtered | |
IL-2 | |
Store in refrigerator (2–8°C). | |
< 0.1 ng/μg | |
Wet Ice | |
Cytokines & Receptors | |
Lyophilized | |
Immunology, Signal Transduction, Stem Cell Research, Angiogenesis, Oncology | |
95 % |
For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.