missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ IL1B Recombinant Rat Protein

Recombinant IL1B is a bioactive protein intended for use in cell culture applications

Brand:  Gibco™ PRC0815

Code : 50

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10059122

  • £289.00 / Each

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

IL-1 is involved in a number of biological process including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture; Mammalian Cell Culture
24494
Sequential Chromatography
25μg
Store at 2° to 8°C.
Recombinant/Ligand
<0.1ng/μg
Cell Culture
E. coli
Lyophilized
17.3kDa
95%
Rat
Carrier Protein
Interleukins
IL-1β
Cytokines & Receptors
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.