Gibco™ IL1B Recombinant Rat Protein

Recombinant IL1B is a bioactive protein intended for use in cell culture applications

Overview
Brand: Gibco™

Manufacturer Part Number: PRC0814

UNSPSC: 12352202

Code: 50

Additional Details:
Additional Details: Weight: 0.50000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10631114

Quantity Price
1 £ 140.0 / Each
EU Stock 12
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Storage Requirements Store at 2° to 8°C.
Source Rat
Quantity 10μg
Endotoxin Concentration <0.1ng/μg
Expression System E. coli
Purity 95%
Gene ID (Entrez) 24494
Protein Subtype Interleukins
Format Lyophilized
Structural Form Recombinant/Ligand
Protein Family Cytokines & Receptors
Without Additives Carrier Protein
Purification Method Sequential Chromatography
Gene Alias IL-1β
For Use With (Application) Cell Culture; Mammalian Cell Culture
Validated Application Cell Culture
Molecular Weight 17.3kDa

IL-1 is involved in a number of biological process including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.

Shipping Condition: Wet Ice. This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.