missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human IL-1 beta Recombinant Protein

Recombinant Human Interleukin 1, beta (IL1B) is a bioactive protein intended for use in cell culture applications

£72.25 - £4440.00

Specifications

For Use With (Application) Cell Culture
Molecular Weight (g/mol) 17.5 kDa
Purification Method Sequential Chromatography
Gene Alias IL-1β
Species Human
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10394403
View Documents Promotion Details
Gibco™
PHC0814
2μg
£72.25
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10532635
View Documents Promotion Details
Gibco™
PHC0815
10μg
£516.00
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10600934
View Documents Promotion Details
Gibco™
PHC0816
25μg
£1032.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10415533
View Documents Promotion Details
Gibco™
PHC0811
100μg
£2570.00
Each
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
10111113
View Documents Promotion Details
Gibco™
PHC0813
1mg
£4440.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 30-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Human IL-1 beta Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

IL-1 is involved in a number of biological processes including the stimulation of T-helper cells, promotion proliferation and synthesis of immunoglobulins, and the activation of fibroblasts and NK cells.
  • High purity: no interference from other proteins or contaminants
  • High biological activity: more results with less protein
  • Proven compatibility: Gibco proteins bioassayed with Gibco media
  • High Purity Recombinant Bioactive Protein: to help ensure Gibco growth factors are of the highest quality, each protein is analyzed for purity along with structural homogeneity to ensure a biologically active protein
  • High Biological Activity: in-house activity testing of Gibco Recombinant Proteins includes cell proliferation, cytotoxicity, chemotaxis, calcium flux, secondary cytokine up-regulation, induction of surface antigen expression, anti-viral, and protease assays; specific activity is determined by the dose dependent proliferation of murine D10S cells
  • Proven Compatibility: Gibco proteins are bioassayed with Gibco media

Shipping Condition: Wet Ice.

Specifications

Specifications

Cell Culture
Sequential Chromatography
Human
Carrier-Free
E. coli
Recombinant, Ligand
Interleukins
95 %
17.5 kDa
IL-1β
Store in refrigerator (2–8°C).
< 0.1 ng/μg
Cytokines & Receptors
Lyophilized
Gibco™
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate

For Research Use Only. Not intended for animal or human diagnostic or therapeutic use.